missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12201-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.Specifications
ABHD5 | |
Polyclonal | |
Unconjugated | |
ABHD5 | |
ABHD5, CDS, CGI58, IECN2, NCIE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51099, 67469 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WTS1, Q9DBL9 | |
ABHD5 | |
ABHD5 Fusion Protein Ag2840 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction