missing translation for 'onlineSavingsMsg'
Learn More

ABH1 Rabbit anti-Human, Mouse, Rat, Clone: S04-4C5, Novus Biologicals™

Rabbit Monoclonal Antibody

2690.00 NOK - 5435.00 NOK

Specifications

Antigen ABH1
Clone S04-4C5
Applications Western Blot, ELISA
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18329344
View Documents
Bio-Techne
NBP3-14988-25UL
25 μg
2690.00 NOK
25µL
Please to purchase this item. Need a web account? Register with us today!
18397624
View Documents
Bio-Techne
NBP3-14988-100UL
100 μg
5435.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ABH1 Monoclonal antibody specifically detects ABH1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications

Specifications

ABH1
Western Blot, ELISA
Unconjugated
Rabbit
DNA Repair
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
8846
IgG
Affinity-purified
S04-4C5
Monoclonal
Purified
RUO
Human, Mouse, Rat
ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH
Recombinant protein of human ABH1 (Uniprot#: Q13686)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.