missing translation for 'onlineSavingsMsg'
Learn More

Abamectin, Thermo Scientific Chemicals

Interacts with GABA receptors to cause chloride channel activation

1240.00 NOK - 3885.00 NOK

Chemical Identifiers

CAS 71751-41-2
Molecular Formula C48H72O14
MDL Number MFCD01769550
Synonym Avermectin B1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472879
View Documents
Thermo Scientific Alfa Aesar
J60039.03
1 g
1240.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
15482879
View Documents
Thermo Scientific Alfa Aesar
J60039.06
5 g
3885.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

Solubility
Soluble in DMSO

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

71751-41-2
C48H72O14
MFCD01769550
Avermectin B1

Specifications

71751-41-2
White to Yellow
MFCD01769550
14,2
Soluble in DMSO
Powder
150°C to 155°C
C48H72O14
UN2811
Avermectin B1
873.1
Abamectin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H372-H361
Fatal if swallowed.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H300+H330-H361d-H372

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'rtecsNumber' : CL1203000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only