missing translation for 'onlineSavingsMsg'
Learn More

AAGAB Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen AAGAB
Concentration 0.5 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16845565
View Documents Promotion Details
Proteintech
27319-1-AP-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16835565
View Documents Promotion Details
Proteintech
27319-1-AP-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The predicted MW of this protein is 35 kDa. 27319-1-AP recognizes a 37-40 kDa band due to phosphorylation.

The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

AAGAB
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
Q6PD74, Q8R2R3
66939, 79719
AAGAB Fusion Protein Ag26340
Primary
-20°C
AAGAB
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
AAGAB, FLJ11506
Aagab
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications