Learn More
Tocris Bioscience™ A01
High affinity Smurf1 inhibitor; enhances BMP signaling
Brand:  Tocris Bioscience™ 5397/50
7188.58 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18767078
  • 9080.00 NOK / 50mg
Explore more special offers
Description
Description
A01 is a high affinity Smurf1 E3 ubiquitin ligase inhibitor (Kd = 3.7 nM). Attenuates Smurf1-mediated Smad 1/5 degradation and enhances BMP signaling. Potentiates BMP-2-induced osteoblastic activity in C2C12 myoblasts and MC3T3-E1 osteoblast precursor cells.
Specifications
Specifications
>98%
A01
A01
50 mg
512.93
Special Offers
For Research Use Only