Learn More
IKA™ A 11.6 Double Beater
Surface-hardened titanium beater with two blades for use in a grinding chamber.
Brand: IKA™ 0003302900
This item is not returnable.
View return policy
Description
- For use with items up to Mohs hardness 3.
- Only applicable with grinding chamber A 11.4.
- Surface-hardened titanium construction.
Specifications
Double Beaker | |
Materials with Mohs hardness up to 3 | |
A 11.4 Grinding chamber | |
Surface-hardened Titanium |
Product Content Correction
Product Title
IKA™ A 11.6 Double Beater > For use with: A 11.4 Grinding chamber