Learn More
Ratiolab™ 96-Well Polystyrene Micro Test Plates
With a precise design, perfectly shaped for use with automatic systems.
665.00 NOK
Specifications
Barcode Alphanumeric
Format SBS Standard
Array 8 x 12
Color Clear
For Use With (Equipment) Automatic systems
View More Specs
Products 2
Product Code Brand Well Shape Price Quantity & Availability  
Product Code Brand Well Shape Price Quantity & Availability  
11315872
View Documents Promotion Details
Ratiolab™
6018111
Round Bottom (U-well)
665.00 NOK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
11971401
View Documents Promotion Details
Ratiolab™
6018311
Conical Bottom (V-well)
665.00 NOK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-11-2024
Description
  • Manufactured from clear polystyrene
  • 96 well with volume approx. 300 μL
  • With U- or V-bottom
  • SBS standard micro test plate format
  • 8 x 12 wells alphanumerically coded
  • Wells with raised rims for reduced cross-contamination and effective sealing with Ratiolab cover films
  • Optimum space for precise positioning
  • Space-saving and stackable
  • Provided in packs of 100
Specifications
Specifications
Alphanumeric
8 x 12
Automatic systems
96
100/Pk.
SBS Standard
Clear
Polystyrene
Non-sterile
Raised