Learn More
QuikPrep 96-Well Equilibrium Dialyzer
Description
The QuikPrep™ 96-Well DispoEquilibrium Dialyzer™ is a unique product for simultaneous assay of 96 samples. Each well has separate membrane eliminating the chance of cross contamination and can hold samples volumes between 50 and 200 μL.
The plate is available with 10 kDa regenerated cellulose membranes. The wells can be sealed with either a pierceable self-sealing mat or through eight cap strips which allow you to use the plate in sections, saving unneeded wells for future use. Each plate come with 24 eight cap strips.
96-Well DispoEquilibrium Dialyzer Plates must be used with a plate rotator. The rotator increases the rate of dialysis and allows for constant motion of samples ensuring higher reproducibility. For temperature controlled studies the 8-Plate Rotator Incubator can be used.
Key Features
- 96-well format
- Individual membrane for each well
- Small sample volumes: 50 μl to 200 μl
- Ultra-thin regenerated cellulose membranes
- Membranes are free of sulfur and heavy metal contamination
- High well-to-well reproducibility
- Excellent sample recovery (>95%)
- Patented
Applications
- Protein and protein-drug binding assays
- Receptor binding assays
- Ligand binding assays
- Protein-protein and Protein-DNA interactions
Specifications
Specifications
Format | Plate |
Membrane | Regenerated Cellulose, Dialysis |
MWCO | 10 kDa |