Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
Brand: Corning™ 3904
10420.22 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Standard | |
0.32 cm2 | |
20/Pk., 100/Cs. | |
Assay Plate | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
With Lid | |
8 x 12 | |
Black, Clear Bottom | |
Polystyrene | |
75 to 200 ÎĽL | |
Cell Culture | |
Attachment and growth of anchorage-dependent cells | |
20/Pk. |
COR001 Lifescience Guid
File size: 7 MB
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Product Content Correction
Product Title
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate > Black and Clear; Standard, with Lid; Yes; TC-treated; Flat