Learn More
uvex 9192 Pheos Safety Glasses
Shop All Uvex Products
Click to view available options
Frame Color:
Black, Gray
Black, Orange
Blue, Gray
Lens Tint:
Clear
Gray
Description
- Perfect all-round protection, with a large field of vision through duo spherical lens
- Outstanding ventilation thanks to integrated uvex climazone temple concept
- Supravision excellence coating is anti-fog on the inside, and extremely scratch-resistant and chemical-resistant on the outside
Specifications
Specifications
Lens Type | Supravision Excellence |
Lens Coating | Anti-fog |
Certifications/Compliance (Europe) | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
Lens Tint | Gray |
Product Type | Ergonomic Safety Glasses |
Frame Color | Black, Gray |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Plastic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
uvex 9192 Pheos Safety Glasses >