missing translation for 'onlineSavingsMsg'
Learn More
9,10-Dibromoanthracene, 98% Product Code.: 11303316

9,10-Dibromoanthracene, 98%

Product Code. 11303316
100 g, 100g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11303316

Brand: Thermo Scientific Alfa Aesar L12680.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 523-27-3 | C14H8Br2 | 336.026 g/mol

9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-27-3
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.026
MDL Number MFCD00001244
InChI Key BRUOAURMAFDGLP-UHFFFAOYSA-N
Synonym anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y
PubChem CID 68226
IUPAC Name 9,10-dibromoanthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br

Specifications

Melting Point 222°C to 223°C (sublimation)
Quantity 100 g
Beilstein 1912109
Merck Index 14,3018
Solubility Information Soluble in water.
Formula Weight 336.04
Percent Purity 98%
Chemical Name or Material 9,10-Dibromoanthracene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9,10-Dibromoanthracene, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.