missing translation for 'onlineSavingsMsg'
Learn More

7-Oxabicyclo[2.2.1]heptane, 98%, Thermo Scientific Chemicals

Product Code. 11425207
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 279-49-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00082342
InChI Key YPWFNLSXQIGJCK-UHFFFAOYSA-N
Synonym 1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
PubChem CID 9234
IUPAC Name 7-oxabicyclo[2.2.1]heptane
SMILES C1CC2CCC1O2

Specifications

Density 0.968
Boiling Point 118°C to 120°C (713 mmHg)
Quantity 5 g
Refractive Index 1.448
UN Number UN3271
Formula Weight 98.15
Percent Purity 98%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'