missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%, Thermo Scientific Chemicals

1683.39 NOK - 8748.53 NOK

Chemical Identifiers

CAS 799274-06-9
Molecular Formula C9H10FN·ClH
Molecular Weight (g/mol) 187.64
InChI Key LOJNQXIJLCPQDR-UHFFFAOYSA-N
Synonym 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-fluoro-1,2,3,4-tetrahydroisoquinoline hcl, 7-fluoro-1,2,3,4-tetrahydroisoquinolinehydrochloride, 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride salt, isoquinoline, 7-fluoro-1,2,3,4-tetrahydro-, hydrochloride, 7-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride salt, 7-fluoro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 45073971
IUPAC Name 7-fluoro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)F.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15259686
View Documents
Thermo Scientific Acros
450460010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15259656
View Documents
Thermo Scientific Acros
450460050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.