missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

795.00 NOK - 6375.00 NOK

Chemical Identifiers

CAS 554405-17-3
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
InChI Key SAZSDHZXRMXESN-UHFFFAOYSA-N
Synonym 6-morpholinopyridine-2-carboxylic acid, 6-morpholinopicolinic acid, 6-morpholin-4-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-4-morpholinyl, 6-morpholinopyridine-2-carboxylicacid, 6-morpholin-4-yl picolinic acid, 6-4-morpholinyl-2-pyridinecarboxylic acid
PubChem CID 2388921
IUPAC Name 6-morpholin-4-ylpyridine-2-carboxylic acid
SMILES C1COCCN1C2=CC=CC(=N2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10113424
SDS View Documents
Thermo Scientific Maybridge
CC38301CB
250 mg Amber Glass Bottle
795.00 NOK
250mg
Please to purchase this item. Need a web account? Register with us today!
10077382
SDS View Documents
Thermo Scientific Maybridge
CC38301DA
1 g Amber Glass Bottle
2230.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
10431334
SDS View Documents
Thermo Scientific Maybridge
CC38301DE
5 g Amber Glass Bottle
6375.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Morpholinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.