missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals

719.00 NOK - 6950.00 NOK

Chemical Identifiers

CAS 3453-33-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00081152
InChI Key VZBLASFLFFMMCM-UHFFFAOYSA-N
Synonym 6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395
PubChem CID 76991
IUPAC Name 6-methoxynaphthalene-2-carbaldehyde
SMILES COC1=CC=C2C=C(C=O)C=CC2=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11456816
SDS View Documents
Thermo Scientific Alfa Aesar
B23943.06
5 g
719.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
11466816
SDS View Documents
Thermo Scientific Alfa Aesar
B23943.14
25 g
2150.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
11476816
SDS View Documents
Thermo Scientific Alfa Aesar
B23943.22
100 g
6950.00 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 222-377-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.