missing translation for 'onlineSavingsMsg'
Learn More
6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals Product Code.: 11466816

6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11466816
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11466816

Brand: Thermo Scientific Alfa Aesar B23943.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3453-33-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00081152
InChI Key VZBLASFLFFMMCM-UHFFFAOYSA-N
Synonym 6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395Show MoreShow Less
PubChem CID 76991
IUPAC Name 6-methoxynaphthalene-2-carbaldehyde
SMILES COC1=CC=C2C=C(C=O)C=CC2=C1

Specifications

Melting Point 80°C to 82°C
Quantity 25 g
Beilstein 1945989
Sensitivity Air Sensitive
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 6-Methoxy-2-naphthaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10231673 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberB23943.06,B23943.14
Lot Number10190106 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23943.06,B23943.14
Lot Number10100206 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23943.06,B23943.14
Lot Number10225963 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23943.06,B23943.14
Lot Number10049474 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23943.06,B23943.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.