Learn More
6,7-Dimethoxy-1-tetralone, 97%
CAS: 13575-75-2 | C12H14O3 | 206.24 g/mol
778.00 NOK - 11025.00 NOK
Chemical Identifiers
CAS | 13575-75-2 |
---|---|
Molecular Formula | C12H14O3 |
Molecular Weight (g/mol) | 206.24 |
MDL Number | MFCD00134100 |
InChI Key | YNNJHKOXXBIJKK-UHFFFAOYSA-N |
Synonym | 6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843 |
PubChem CID | 266816 |
IUPAC Name | 6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one |
SMILES | COC1=CC2=C(C=C1OC)C(=O)CCC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480738
|
Thermo Scientific Alfa Aesar
B25226.03 |
1 g |
778.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490738
|
Thermo Scientific Alfa Aesar
B25226.06 |
5 g |
2345.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400748
|
Thermo Scientific Alfa Aesar
B25226.14 |
25 g |
11025.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13575-75-2 | |
206.24 | |
YNNJHKOXXBIJKK-UHFFFAOYSA-N | |
266816 | |
COC1=CC2=C(C=C1OC)C(=O)CCC2 |
C12H14O3 | |
MFCD00134100 | |
6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843 | |
6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one |
Specifications
13575-75-2 | |
C12H14O3 | |
6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843 | |
COC1=CC2=C(C=C1OC)C(=O)CCC2 | |
206.24 | |
206.24 | |
6,7-Dimethoxy-1-tetralone |
98°C to 100°C | |
MFCD00134100 | |
YNNJHKOXXBIJKK-UHFFFAOYSA-N | |
6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one | |
266816 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.