Learn More
6,7-Dimethoxy-1-tetralone, 97%, Thermo Scientific Chemicals Product Code.: 11400748
6,7-Dimethoxy-1-tetralone, 97%, Thermo Scientific Chemicals
Product Code. 11400748
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
Product Code. 11400748
Brand: Thermo Scientific Alfa Aesar B25226.14
Request Bulk or Custom Format
CAS: 13575-75-2 | C12H14O3 | 206.24 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13575-75-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00134100
InChI Key YNNJHKOXXBIJKK-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843
PubChem CID 266816
IUPAC Name 6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(C=C1OC)C(=O)CCC2
Specifications
Melting Point 98°C to 100°C
Quantity 25 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 6,7-Dimethoxy-1-tetralone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10232438 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25226.03,B25226.14,B25226.06
Lot Number10177232 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25226.03,B25226.14,B25226.06
Lot Number10154403 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25226.03,B25226.14,B25226.06
Product Title
6,7-Dimethoxy-1-tetralone, 97%, Thermo Scientific Chemicals >