missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pergolide mesylate, MedChemExpress

Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
Pergolide mesylate (Pergolide methanesulfonate), an Ergoline derivative, is a potent and orally active dopamine D1 and D2 receptors agonist. Pergolide mesylate can be used for Parkinson's disease and hyperprolactinaemia research.
Specifications
Specifications
Chemical Name or Material | Pergolide mesylate |
CAS | 66104-23-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C20H30N2O3S2 |
Quantity | 50 mg |
Synonym | Pergolide methanesulfonate LY127809 |
Solubility Information | DMSO : ≥ 25 mg/mL (60.89 mM) |
SMILES | [H][C@@]1(N(CCC)C[C@H](CSC)C[C@@]12[H])CC3=CNC4=CC=CC2=C43.CS(=O)(O)=O |
Molecular Weight (g/mol) | 410.59 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Pergolide mesylate, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction