missing translation for 'onlineSavingsMsg'
Learn More

5-Undecanol, 98%, Thermo Scientific™

Product Code. 11326756
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37493-70-2
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00046719
InChI Key PCFOZHPCKQPZCN-UHFFFAOYNA-N
Synonym 5-undecanol, undecanol-5, 5-undecanol, s, acmc-20mbit, acmc-1ct1g
PubChem CID 98676
IUPAC Name undecan-5-ol
SMILES CCCCCCC(O)CCCC

Specifications

Melting Point -4°C
Density 0.83
Boiling Point 139°C to 140°C (17 mmHg)
Refractive Index 1.4362
Quantity 1 g
Beilstein 1698382
Formula Weight 172.31
Percent Purity 98%
Chemical Name or Material 5-Undecanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'