missing translation for 'onlineSavingsMsg'
Learn More
5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+% Product Code.: 15458117

5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%

Product Code. 15458117
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15458117

Brand: Thermo Scientific Alfa Aesar H51531.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 848613-91-2 | C16H31NSSn | 388.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 848613-91-2
Molecular Formula C16H31NSSn
Molecular Weight (g/mol) 388.201
MDL Number MFCD09025809
InChI Key LALGELPHLJBAEK-UHFFFAOYSA-N
Synonym 5-methyl-2-tributylstannyl thiazole, 5-methyl-2-tributylstannyl-1,3-thiazole, 5-methyl-2-tri-n-butylstannyl thiazole, acmc-209px3, 5-methyl-2-tributylstannyl-thiazole, thiazole, 5-methyl-2-tributylstannyl, 5-methyl-2-tributylstannanyl-thiazole, 5-methyl-2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl-5-methylthiazole
PubChem CID 16427131
IUPAC Name tributyl-(5-methyl-1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=C(S1)C

Specifications

Quantity 1 g
UN Number UN2788
Formula Weight 388.19
Percent Purity ≥90%
Chemical Name or Material 5-Methyl-2-(tri-n-butylstannyl)thiazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot NumberU22D004 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51531.MD,H51531.03
Lot NumberM23E054 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51531.MD,H51531.03
Lot NumberX13B003 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51531.MD,H51531.03
Lot NumberB04X040 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51531.MD,H51531.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.