missing translation for 'onlineSavingsMsg'
Learn More
5-Chlorothiophene-2-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11422228

5-Chlorothiophene-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11422228
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11422228

Brand: Thermo Scientific Alfa Aesar A15209.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24065-33-6
Molecular Formula C5H3ClO2S
Molecular Weight (g/mol) 162.59
MDL Number MFCD00041426
InChI Key QZLSBOVWPHXCLT-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2jShow MoreShow Less
PubChem CID 95048
IUPAC Name 5-chlorothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 149°C to 152°C
Quantity 25 g
Beilstein 118361
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 5-Chlorothiophene-2-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10164231 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA15209.06,A15209.22,A15209.14
Lot NumberL10T007 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15209.06,A15209.22,A15209.14
Lot NumberE08T006 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15209.06,A15209.22,A15209.14
Lot NumberI4103A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15209.06,A15209.22,A15209.14
Lot Number5006A30W Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15209.06,A15209.22,A15209.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chlorothiophene-2-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.