missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosulfonyl-2-methoxybenzoic acid, 99%, Thermo Scientific™

Product Code. p-7033284
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51904-91-7
Molecular Formula C8H7ClO5S
Molecular Weight (g/mol) 250.649
MDL Number MFCD00066912
InChI Key OUBZCDOQEMLMAB-UHFFFAOYSA-N
Synonym 5-chlorosulfonyl-2-methoxybenzoic acid, 5-chlorosulphonyl-2-anisic acid, benzoic acid, 5-chlorosulfonyl-2-methoxy, 5-chlorosulphonyl-2-methoxybenzoic acid, 5-chlorosulfonyl-2-methoxybenzoicacid, acmc-1ar14, 2-methoxy-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-methoxy-benzoic acid, 5-chlorosulphonyl-2-methoxy benzoic acid, 5-chlorosulphonyl-2-methoxy-benzoic acid
PubChem CID 104007
IUPAC Name 5-chlorosulfonyl-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(=O)O

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 250.66
Percent Purity 99%
Chemical Name or Material 5-Chlorosulfonyl-2-methoxybenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chlorosulfonyl-2-methoxybenzoic acid, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.