missing translation for 'onlineSavingsMsg'
Learn More
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™ Product Code.: 15491176

5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15491176
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15491176

Brand: Thermo Scientific Alfa Aesar H54432.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1083168-91-5
Molecular Formula C12H17BClNO3
Molecular Weight (g/mol) 269.532
MDL Number MFCD08063054
InChI Key ZHWIZWNYOSGXSR-UHFFFAOYSA-N
Synonym 3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-chloro-6-methoxypyridine-3-boronic acid pinacol ester, 3-chloro-2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxypyridine-5-boronic acid pinacol ester, 5-chloro-6-methoxypyridin-3-yl boronic acid pinacol ester, 3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pyridine, 3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 53398545
IUPAC Name 3-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)Cl

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 269.53
Percent Purity 96%
Chemical Name or Material 5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.