missing translation for 'onlineSavingsMsg'
Learn More
5-Chloro-2-fluoropyridine-3-boronic acid, 97%, Thermo Scientific™ Product Code.: 15400088

5-Chloro-2-fluoropyridine-3-boronic acid, 97%, Thermo Scientific™

Product Code. 15400088
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15400088

Brand: Thermo Scientific Alfa Aesar H53341.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937595-70-5
Molecular Formula C5H4BClFNO2
Molecular Weight (g/mol) 175.35
MDL Number MFCD04972408
InChI Key ZUDXVHDVUTUTFH-UHFFFAOYSA-N
Synonym 5-chloro-2-fluoropyridine-3-boronic acid, 5-chloro-2-fluoropyridin-3-yl boronic acid, 5-chloro-2-fluoropyridin-3-ylboronicacid, 2-fluoro-5-chloropyridine-3-boronic acid, 5-chloro-2-fluoro-3-pyridyl boronic acid, 5-chloro-2-fluoropyridine-3-boronic acid hydrate, pubchem17095, acmc-209rnf, 5-chloro-2-fluoro-3-pyridylboronic acid, 5-chloro-2-fluoro-3-pyridinyl boronic acid
PubChem CID 24901779
IUPAC Name (5-chloro-2-fluoropyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1F)Cl)(O)O

Specifications

Melting Point 125°C to 126°C
Quantity 1 g
Formula Weight 175.35
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluoropyridine-3-boronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot NumberBK060501 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chloro-2-fluoropyridine-3-boronic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.