missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxamidotryptamine maleate, Tocris Bioscience™

Product Code. 30000699
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT1 agonist; also has high affinity for 5-ht5a and 5-HT7

5-HT1 agonist with high affinity at 5-HT1A, 5-HT1B, 5-HT1D, 5-ht5 and 5-HT7 receptors.

Chemical Identifiers

CAS 74885-72-6
Molecular Formula C11H13N3O·C4H4O4

Specifications

Quantity 10 mg
Formula Weight 319.32
Percent Purity >99%
Chemical Name or Material 5-Carboxamidotryptamine maleate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'