missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%

CAS: 205926-98-3 | C8H4BrClNNa2O4P | 370.43 g/mol

502.00 NOK - 4400.00 NOK

Chemical Identifiers

CAS 205926-98-3
Molecular Formula C8H4BrClNNa2O4P
Molecular Weight (g/mol) 370.43
MDL Number MFCD00150468
InChI Key OAZUOCJOEUNDEK-UHFFFAOYSA-L
PubChem CID 6097197
IUPAC Name disodium;(5-bromo-4-chloro-1H-indol-3-yl) phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10301374
SDS View Documents
Thermo Scientific Acros
271460500
50 mg Glass bottle
502.00 NOK
50mg
Please to purchase this item. Need a web account? Register with us today!
10638391
SDS View Documents
Thermo Scientific Acros
271462500
250 mg Glass bottle
1545.00 NOK
250mg
Please to purchase this item. Need a web account? Register with us today!
10530974
SDS View Documents
Thermo Scientific Acros
271460010
1 g Glass bottle
4400.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.