missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylindole, 96%

Product Code. 11976661 Shop All Thermo Scientific Chemicals Products
5 g, Glass bottle, 5g
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1075-34-9 | C9H8BrN | 210.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-34-9
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.07
InChI Key BJUZAZKEDCDGRW-UHFFFAOYSA-N
PubChem CID 5003968
IUPAC Name 5-bromo-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 104.0°C to 106.0°C
Color Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Synonym 5-bromo-2-methylindole, 2-methyl-5-bromoindole, 1h-indole, 5-bromo-2-methyl, 5-bromo-2-methylindol, pubchem9382, acmc-209xly, 5-bromo-2-methyl-indole, 2-methyl-5-bromo-1h-indole, 1h-indole,5-bromo-2-methyl
Formula Weight 210.07
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-2-methylindole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-2-methylindole, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.