missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-2'-Deoxyuridine, MP Biomedicals™ Product Code.: 11338489
compliance-icons

5-Bromo-2'-Deoxyuridine, MP Biomedicals™

Product Code. 11338489 Shop All MP Biomedicals Products
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 11338489

Brand: MP Biomedicals 02100171.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful for the study of DNA synthesis. 5-Bromo-2'-Deoxyuridine, MP Biomedicals (5-BrdU) is a thymidine analogue that is incorporated into DNA. It is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently it is us

5-BrdU can be used to label DNA, which can be used in conjunction with BrdU Monoclonal Antibody for the immunohistochemical analysis of cell proliferation. Thymidine analog is used as a mutagen in genetic research. It is selectively incorporated into cellular DNA in place of thymidine during S-phase. It is also used to study cell signaling and other processes that induce cell proliferation.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Specifications

Melting Point 183°C to 193°C
Quantity 500 mg
Solubility Information Soluble in water (10mg/mL or 50–100mg/mL with heat) or sodium hydroxide (50mg/mL).
Formula Weight 307.1
Percent Purity ≥99%
Physical Form Powder
compliance-icons
Product Identifier
  • 5-BROMO-2'-DEOXYURIDINE   500MG
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-2'-Deoxyuridine, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.