missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxycytidine, MP Biomedicals™

Product Code. 11398479
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
250 mg
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1022-79-3
Molecular Formula C9H12BrN3O4
Molecular Weight (g/mol) 306.116
InChI Key KISUPFXQEHWGAR-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy
PubChem CID 472175
IUPAC Name 4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O

Specifications

Color White
Quantity 250 mg
Formula Weight 306.1 (Anhydrous)
Physical Form Powder
Chemical Name or Material 5-Bromo-2'-deoxycytidine monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'