missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2'-deoxycytidine, MP Biomedicals™
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
Specifications
Specifications
Color | White |
Quantity | 100 mg |
Formula Weight | 306.1 (Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | 5-Bromo-2'-deoxycytidine monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'