missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6,7,8-Tetrahydro-1-naphthylamine, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 38.0°C |
Color | Yellow to Brown |
Density | 1.0560g/mL |
Boiling Point | 275.0°C to 277.0°C (713.0 mmHg) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling

Product Identifier
- 5,6,7,8-Tetrahydro-1-naphthylamine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5,6,7,8-Tetrahydro-1-naphthylamine, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction