Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve with Square Holes, ISO Certified
Brand: RETSCH 60.154.080000
This item is not returnable.
View return policy
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Stainless Steel |
80 mm | |
ISO 3310/2 | |
65mm | |
400 mm | |
Stainless Steel |
Product Content Correction
Product Title
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve with Square Holes, ISO Certified > Pore Size: 80 mm