missing translation for 'onlineSavingsMsg'
Learn More
4-Trifluoromethyl-L-phenylalanine, 95% Product Code.: 15488707

4-Trifluoromethyl-L-phenylalanine, 95%

Product Code. 15488707
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15488707

Brand: Thermo Scientific Alfa Aesar H52170.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114926-38-4 | C10H10F3NO2 | 233.19 g/mol

4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114926-38-4
Molecular Formula C10H10F3NO2
Molecular Weight (g/mol) 233.19
MDL Number MFCD00044946
InChI Key CRFFPDBJLGAGQL-QMMMGPOBSA-N
Synonym 4-trifluoromethyl-l-phenylalanine, s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, l-4-trifluoromethylphenylalanine, l-4-trifluoromethylphe, l-phenylalanine, 4-trifluoromethyl, 4-trifluromethyl-l-phenylalanine hydrochloride, s-2-amino-3-4-trifluoromethyl phenyl propionic acid, h-phe 4-cf3-ohShow MoreShow Less
PubChem CID 2761501
IUPAC Name (2S)-2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)C(F)(F)F

Specifications

Beilstein 5747857
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 233.19
Percent Purity 95%
Chemical Name or Material 4-Trifluoromethyl-L-phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberS13K018 Certificate TypeCertificate of Analysis Date28/06/2024 Product CodeH52170.MD,H52170.03,H52170.06
Lot NumberM25K017 Certificate TypeCertificate of Analysis Date01/02/2024 Product CodeH52170.MD,H52170.03,H52170.06
Lot NumberM26J009 Certificate TypeCertificate of Analysis Date24/10/2023 Product CodeH52170.MD,H52170.03,H52170.06
Lot NumberT19J019 Certificate TypeCertificate of Analysis Date25/07/2023 Product CodeH52170.MD,H52170.03,H52170.06
Lot NumberX26I012 Certificate TypeCertificate of Analysis Date18/01/2023 Product CodeH52170.MD,H52170.03,H52170.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Trifluoromethyl-L-phenylalanine, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.