Learn More
4-(Trifluoromethoxy)phenol, 98%, Thermo Scientific™
Product Code. 11460603
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 828-27-3
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040988
InChI Key WDRJNKMAZMEYOF-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol
PubChem CID 70015
IUPAC Name 4-(trifluoromethoxy)phenol
SMILES OC1=CC=C(OC(F)(F)F)C=C1
Specifications
Melting Point 17°C to 18°C
Density 1.375
Boiling Point 91°C to 92°C (25 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.447
Quantity 5 g
UN Number UN2810
Beilstein 1945934
Sensitivity Hygroscopic
Formula Weight 178.11
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)phenol