missing translation for 'onlineSavingsMsg'
Learn More
4-tert-Butylphenol, 97% Product Code.: 10657632

4-tert-Butylphenol, 97%

Product Code. p-80842 Shop All Thermo Scientific Chemicals Products
5 g, 5g
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
Unit Size:
1kg
2.50kg
5g
This item is not returnable. View return policy

Product Code. 10657632

Brand: Thermo Scientific Acros 108000050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-54-4 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(C=C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 96.0°C to 100.0°C
Color White to Beige
Density 0.9000g/mL
Boiling Point 236.0°C to 238.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4OH
Refractive Index 1.4787
Quantity 5 g
Beilstein 06, 524
Merck Index 15, 1588
Specific Gravity 0.9
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in aq. alcoholic alkali,soluble in alcohol and ether
Viscosity 2.4 mPa.s (110°C)
Formula Weight 150.22
Percent Purity 97%
Physical Form Flakes or Pastilles
Chemical Name or Material 4-tert-Butylphenol, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-tert-Butylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-tert-Butylphenol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.