missing translation for 'onlineSavingsMsg'
Learn More
4-(tert-Butyl)benzene-1-carbothioamide, ≥95%, Thermo Scientific™ Product Code.: 10702693

4-(tert-Butyl)benzene-1-carbothioamide, ≥95%, Thermo Scientific™

Product Code. 10702693
10 g
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 10702693

Brand: Thermo Scientific Maybridge RF00754EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57774-77-3
Molecular Formula C11H15NS
Molecular Weight (g/mol) 193.308
MDL Number MFCD00052485
InChI Key HZODWYBXBKXJLB-UHFFFAOYSA-N
Synonym 4-tert-butyl benzothioamide, 4-tert-butylthiobenzamide, 4-tert-butylbenzene-1-carbothioamide, 4-tert-butyl-thiobenzamide, 4-tert-butyl benzene-1-carbothioamide, amino 4-tert-butyl phenyl methane-1-thione, acmc-1az3y, benzenecarbothioamide,4-1,1-dimethylethyl, benzenecarbothioamide, 4-1,1-dimethylethyl-9ci
PubChem CID 2734817
IUPAC Name 4-tert-butylbenzenecarbothioamide
SMILES CC(C)(C)C1=CC=C(C=C1)C(=S)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula (CH3)3CC6H4CSNH2
Quantity 10 g
Formula Weight 193.31
Percent Purity 95%
Chemical Name or Material 4-(tert-Butyl)benzene-1-carbothioamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(tert-Butyl)benzene-1-carbothioamide, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.