missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals Product Code.: 11343156

4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11343156
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11343156

Brand: Thermo Scientific Alfa Aesar L11042.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

Solubility
It hydrolyzes with water. Soluble in toluene.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-61-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD00007307
InChI Key NXHSSIGRWJENBH-UHFFFAOYSA-N
Synonym 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanateShow MoreShow Less
PubChem CID 75044
IUPAC Name 1-isothiocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=S)[N+](=O)[O-]

Specifications

Melting Point 105°C to 110°C
Boiling Point 137°C to 138°C (11 mmHg)
Odor Irritating
Quantity 50 g
UN Number UN2923
Beilstein 640027
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water. Soluble in toluene.
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl isothiocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248769 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberL11042.30,L11042.18,L11042.09
Lot Number10216695 Certificate TypeCertificate of Analysis Date22/06/2023 Catalog NumberL11042.30,L11042,L11042.18,L11042.09
Lot Number10243403 Certificate TypeCertificate of Analysis Date09/09/2022 Catalog NumberL11042.30,L11042.18,L11042.09
Lot Number10139881 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11042.30,L11042.18,L11042.09
Lot Number10137326 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11042.30,L11042.18,L11042.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.