Learn More
4-Nitrophenyl acetate, 97%
CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol
317.00 NOK - 2170.00 NOK
Chemical Identifiers
CAS | 830-03-5 |
---|---|
Molecular Formula | C8H7NO4 |
Molecular Weight (g/mol) | 181.15 |
MDL Number | MFCD00007326 |
InChI Key | QAUUDNIGJSLPSX-UHFFFAOYSA-N |
Synonym | p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester |
PubChem CID | 13243 |
ChEBI | CHEBI:82635 |
SMILES | CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10773275
|
Thermo Scientific Acros
128750050 |
5 g | Glass bottle |
317.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10113660
|
Thermo Scientific Acros
128750250 |
25 g | Plastic bottle |
799.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386720
|
Thermo Scientific Acros
128751000 |
100 g | Plastic bottle |
2170.00 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
830-03-5 | |
181.15 | |
QAUUDNIGJSLPSX-UHFFFAOYSA-N | |
13243 | |
CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O |
C8H7NO4 | |
MFCD00007326 | |
p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester | |
CHEBI:82635 |
Specifications
830-03-5 | |
100.0 | |
Green | |
96% min. (GC) | |
CH3CO2C6H4NO2 | |
06, 233 | |
p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester | |
QAUUDNIGJSLPSX-UHFFFAOYSA-N | |
(4-nitrophenyl) acetate | |
13243 | |
181.15 | |
Powder |
96.0 | |
76.0°C to 79.0°C | |
Authentic | |
C8H7NO4 | |
MFCD00007326 | |
02,297 | |
Solubility in water: insoluble | |
CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O | |
181.15 | |
CHEBI:82635 | |
97% | |
4-Nitrophenyl acetate, 97% |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-593-5
missing translation for 'rtecsNumber' : AJ1150000
missing translation for 'tsca' : TSCA
- 4-Nitrophenyl acetate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.