missing translation for 'onlineSavingsMsg'
Learn More
4-(Methylthio)benzonitrile, 98+%, Thermo Scientific™ Product Code.: 11362816

4-(Methylthio)benzonitrile, 98+%, Thermo Scientific™

Product Code. 11362816
100 g, 100g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11362816

Brand: Thermo Scientific Alfa Aesar L08979.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(Methylthio)benzonitrile was used in the preparation of 4-cyanobenzenethiol.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21382-98-9
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00010381
InChI Key RPVGEEHGKIFQFO-UHFFFAOYSA-N
Synonym 4-methylthio benzonitrile, 4-methylsulfanyl benzonitrile, benzonitrile, 4-methylthio, 4-methylthiobenzenecarbonitrile, 4-cyanothioanisole, 4-cyano-thioanisole, pubchem10528, 4-methylthiobenzonitrile, p-methylthio benzonitrile
PubChem CID 590960
IUPAC Name 4-methylsulfanylbenzonitrile
SMILES CSC1=CC=C(C=C1)C#N

Specifications

Melting Point 59°C to 64°C
Boiling Point 272°C
Quantity 100 g
UN Number UN3439
Beilstein 2079565
Solubility Information Insoluble in water.
Formula Weight 149.22
Percent Purity ≥98%
Chemical Name or Material 4-(Methylthio)benzonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot Number10145763 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10208877 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10191061 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10190596 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Methylthio)benzonitrile, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.