missing translation for 'onlineSavingsMsg'
Learn More
4-Methyl-5-nonanol, 98% Product Code.: 11410757

4-Methyl-5-nonanol, 98%

Product Code. 11410757
250 g, 250g
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11410757

Brand: Thermo Scientific Alfa Aesar B23167.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 154170-44-2 | C10H22O | 158.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154170-44-2
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.29
MDL Number MFCD00046696
InChI Key MBZNNOPVFZCHID-UHFFFAOYNA-N
Synonym 4-methyl-5-nonanol, 5-nonanol, 4-methyl, 4-methyl 5-nonanol, acmc-1c1h3
PubChem CID 3025865
IUPAC Name 4-methylnonan-5-ol
SMILES CCCCC(O)C(C)CCC

Specifications

Refractive Index 1.436
Quantity 250 g
Formula Weight 158.29
Percent Purity 98%
Chemical Name or Material 4-Methyl-5-nonanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number10168290 Certificate TypeCertificate of Analysis Date24/07/2023 Product CodeB23167.30,B23167.09,B23167.18
Lot Number10168290 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB23167.30,B23167.09,B23167.18
Lot NumberA17X056 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB23167.30,B23167.09,B23167.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyl-5-nonanol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.