missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitrophenol, 99%, Thermo Scientific Chemicals

1034.00 NOK - 5008.00 NOK

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3Show MoreShow Less
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10091521
SDS View Documents
Thermo Scientific Acros
127310250
25 g Glass bottle N/A N/A N/A
10514061
SDS View Documents
Thermo Scientific Acros
127311000
100 g Glass bottle N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyl-2-nitrophenol, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.