Learn More
4-Hexen-3-one, 98%, trans-isomer >95%, Thermo Scientific Chemicals Product Code.: 11317299
4-Hexen-3-one, 98%, trans-isomer >95%, Thermo Scientific Chemicals
Product Code. 11317299
50 g
Click to view available options
Quantity:
10 g
50 g
Product Code. 11317299
Brand: Thermo Scientific Alfa Aesar H27445.18
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2497-21-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00010631
InChI Key FEWIGMWODIRUJM-HYXAFXHYSA-N
Synonym 4-hexen-3-one, 2-hexen-4-one, 2-hexene-4-one, e-hex-4-en-3-one, 4-hexene-3-one, fema no. 3352, e-4-hexen-3-one, 4-hexen-3-one, predominantly trans, e-ch3ch=chc =o c2h5, trans-4-hexen-3-oneShow MoreShow Less
PubChem CID 5365811
IUPAC Name (4Z)-hex-4-en-3-one
SMILES CCC(=O)\C=C/C
Specifications
Density 0.858
Boiling Point 135°C to 137°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.44
Quantity 50 g
UN Number UN1224
Formula Weight 98.15
Percent Purity 98%
Assay trans-isomer >95%
Chemical Name or Material 4-Hexen-3-one
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252897 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog NumberH27445.09,H27445.18
Lot Number10205413 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27445.09,H27445.18
Lot Number10177693 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27445.09,H27445.18
Lot Number10133907 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27445.09,H27445.18
Lot Number10223082 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27445.09,H27445.18
Product Title
4-Hexen-3-one, 98%, trans-isomer >95%, Thermo Scientific Chemicals >