missing translation for 'onlineSavingsMsg'
Learn More
(4-Fluorobenzyl)triphenylphosphonium chloride, 98+%, Thermo Scientific™ Product Code.: 11469822

(4-Fluorobenzyl)triphenylphosphonium chloride, 98+%, Thermo Scientific™

Product Code. 11469822
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11469822

Brand: Thermo Scientific Alfa Aesar A10852.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3462-95-1
Molecular Formula C25H21ClFP
Molecular Weight (g/mol) 406.87
MDL Number MFCD00031637
InChI Key CBHDAHHYMRXLIP-UHFFFAOYSA-M
Synonym 4-fluorobenzyl triphenylphosphonium chloride, phosphonium, 4-fluorophenyl methyl triphenyl-, chloride, 4-fluorophenyl methyl triphenylphosphanium chloride, acmc-20a9k1, ksc495g9t, guljcsointwknf-uhfffaoysa-m, 4-fluorobenzyl triphenylphosphine chloride, p-fluorobenzyltriphenylphosphonium chloride, 4-fluorobenzyltriphenyl phosphonium chloride
PubChem CID 2733554
SMILES [Cl-].FC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 310°C to 312°C
Quantity 5 g
Beilstein 4075331
IUPAC Name [(4-fluorophenyl)methyl]triphenylphosphanium chloride
Formula Weight 406.87
Percent Purity ≥98%
Chemical Name or Material (4-Fluorobenzyl)triphenylphosphonium chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot NumberF8362B Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot NumberF8333A Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4-Fluorobenzyl)triphenylphosphonium chloride, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.