missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-3-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™ Product Code.: 15413557

4-Fluoro-3-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™

Product Code. 15413557
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15413557

Brand: Thermo Scientific Alfa Aesar H26858.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-(trifluoromethoxy)benzylamine are used in natural gas and refinery process streams in industrially amines.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886501-20-8
Molecular Formula C8H7F4NO
Molecular Weight (g/mol) 209.144
MDL Number MFCD06660254
InChI Key LQEZTMITGPWONC-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethoxy phenyl methanamine, 4-fluoro-3-trifluoromethoxy benzylamine, 1-4-fluoro-3-trifluoromethoxy phenyl methanamine, 4-fluoro-3-trifluoromethoxybenzylamine, 4-fluoro-3-trifluoromethoxy phenyl methylamineShow MoreShow Less
PubChem CID 17750702
IUPAC Name [4-fluoro-3-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=C(C=C1CN)OC(F)(F)F)F

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 209.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)benzylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10145474 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10141413 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10170994 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10157072 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-3-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.