missing translation for 'onlineSavingsMsg'
Learn More
4-Ethylphenyl isothiocyanate, 97%, Thermo Scientific Chemicals Product Code.: 11445787

4-Ethylphenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11445787
2 g, 2g
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11445787

Brand: Thermo Scientific Alfa Aesar B24353.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18856-63-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00041101
InChI Key XXLWCGRHJFEMQH-UHFFFAOYSA-N
Synonym 4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
PubChem CID 140438
IUPAC Name 1-ethyl-4-isothiocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=S

Specifications

Boiling Point 245°C
Flash Point >110°C (230°F)
Odor Pungent
Quantity 2 g
UN Number UN2922
Beilstein 2802384
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material 4-Ethylphenyl isothiocyanate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249398 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog NumberB24353.09,B24353.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethylphenyl isothiocyanate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.