missing translation for 'onlineSavingsMsg'
Learn More
4-Ethoxy-2,3-difluorobenzaldehyde, 97%, Thermo Scientific™ Product Code.: 15136725

4-Ethoxy-2,3-difluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 15136725
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15136725

Brand: Thermo Scientific Alfa Aesar H3245003

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Ethoxy-2,3-difluorobenzaldehyde, is used as a pharmaceutical intermediate in antibiotics, as sedatives, and for cancer treatment.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126162-95-6
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.16
MDL Number MFCD09258700
InChI Key DYASUFNACNHJMK-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-ethoxybenzaldehyde, 4-ethoxy-2,3-difluoro-benzaldehyde, 4-ethoxy-2,3-difluoro benzaldehyde
PubChem CID 14495625
IUPAC Name 4-ethoxy-2,3-difluorobenzaldehyde
SMILES CCOC1=C(F)C(F)=C(C=O)C=C1

Specifications

Melting Point 70°C to 73°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 186.16
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10153415 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethoxy-2,3-difluorobenzaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.