missing translation for 'onlineSavingsMsg'
Learn More
4-(Difluoromethoxy)benzyl bromide, 97%, Thermo Scientific™ Product Code.: 11476966

4-(Difluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11476966
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11476966

Brand: Thermo Scientific Alfa Aesar B24052.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store away from incompatible materials such as oxidizing agents. Storage temperature is 2 - 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3447-53-8
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.04
MDL Number MFCD00085007
InChI Key IFNMJBFIRKAQBT-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-difluoromethoxy benzene, 4-difluoromethoxy benzyl bromide, 4-difluoromethoxybenzyl bromide, 4-difluoromethoxy benzylbromide, p-difluoromethoxy benzyl bromide, benzene, 1-bromomethyl-4-difluoromethoxy, 1-bromomethyl-4-difluoromethoxy-benzene, 4-bromomethyl phenoxy difluoromethane, acmc-1aezp, p-difluoromethoxybenzylbromideShow MoreShow Less
PubChem CID 2737001
IUPAC Name 1-(bromomethyl)-4-(difluoromethoxy)benzene
SMILES FC(F)OC1=CC=C(CBr)C=C1

Specifications

Boiling Point 64°C to 66°C (0.3 mmHg)
Odor Pungent
Refractive Index 1.52
Quantity 1 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)benzyl bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10176360 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10183878 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberI2475A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10138384 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberK30W022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Difluoromethoxy)benzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.