Learn More
4-Cyclohexylbenzoic acid, 98%, Thermo Scientific Chemicals
598.00 NOK - 7305.00 NOK
Chemical Identifiers
CAS | 20029-52-1 |
---|---|
Molecular Formula | C13H16O2 |
Molecular Weight (g/mol) | 204.269 |
MDL Number | MFCD00039459 |
InChI Key | QCIWHVKGVVQHIY-UHFFFAOYSA-N |
Synonym | 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexaneShow More |
PubChem CID | 88337 |
IUPAC Name | 4-cyclohexylbenzoic acid |
SMILES | C1CCC(CC1)C2=CC=C(C=C2)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301827
|
Thermo Scientific Alfa Aesar
L05086.04 |
2 g |
598.00 NOK
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311827
|
Thermo Scientific Alfa Aesar
L05086.09 |
10 g |
1820.00 NOK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321827
|
Thermo Scientific Alfa Aesar
L05086.18 |
50 g |
7305.00 NOK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20029-52-1 | |
204.269 | |
QCIWHVKGVVQHIY-UHFFFAOYSA-N | |
88337 | |
C1CCC(CC1)C2=CC=C(C=C2)C(=O)O |
C13H16O2 | |
MFCD00039459 | |
4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexaneShow More | |
4-cyclohexylbenzoic acid |
Specifications
20029-52-1 | |
C13H16O2 | |
1966671 | |
QCIWHVKGVVQHIY-UHFFFAOYSA-N | |
4-cyclohexylbenzoic acid | |
88337 | |
98% |
194°C to 198°C | |
MFCD00039459 | |
4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane | |
C1CCC(CC1)C2=CC=C(C=C2)C(=O)O | |
204.269 | |
204.27 | |
4-Cyclohexylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 243-472-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 4-Cyclohexylbenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 4-Cyclohexylbenzoicacid