missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-4'-n-hexylbiphenyl, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Cyano-4′-n-hexylbiphenyl, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a liquid crystal monomer.
Solubility
Sparingly Soluble in water (3.2E-5 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
4-Cyano-4′-n-hexylbiphenyl, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a liquid crystal monomer.
Solubility
Sparingly Soluble in water (3.2E-5 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
Flash Point | 113°C (235°F) |
Quantity | 250 mg |
Solubility Information | Sparingly Soluble in water (3.2E-5g/L) (25°C). |
Formula Weight | 263.38 |
Percent Purity | 97% |
Chemical Name or Material | 4-Cyano-4'-n-hexylbiphenyl |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction